- Synthesis of Chitosan-graft-Poly (vinyl ether) as a Thermoresponsive Water-Soluble Chitosan
Synthesis of Chitosan-graft-Poly (vinyl ether) as a Thermoresponsive Water-Soluble Chitosan
Journal of Fiber Science and Technology (2019)
Namikoshi T, et al.