Merck
  • Rapid isolation of antigen-specific antibody-secreting cells using a chip-based immunospot array.
Rapid isolation of antigen-specific antibody-secreting cells using a chip-based immunospot array.
Nature protocols (2011-04-30)
Aishun Jin, Tatsuhiko Ozawa, Kazuto Tajiri, Tsutomu Obata, Hiroyuki Kishi, Atsushi Muraguchi
ABSTRACT
Here we report a new method for isolating antigen-specific antibody-secreting cells (ASCs) using a microwell array chip, which offers a rapid, efficient and high-throughput (up to 234,000 individual cells) system for the detection and retrieval of cells that secrete antibodies of interest on a single-cell basis. We arrayed a large population of lymphoid cells containing ASCs from human peripheral blood on microwell array chips and detected spots with secreted antibodies. This protocol can be completed in less than 7 h, including 3 h of cell culture. The method presented here not only has high sensitivity and specificity comparable with enzyme-linked immunospot (ELISPOT) but it also overcomes the limitations of ELISPOT in recovering ASCs that can be used to produce antigen-specific human monoclonal antibodies. This method can also be used to detect cells secreting molecules other than antibodies, such as cytokines, and it provides a tool for cell analysis and clinical diagnosis.
Sigma-Aldrich
Anti-Human IgG (Fc specific)-Cy3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Streptavidin−Cy3 from Streptomyces avidinii, buffered aqueous solution