Merck
  • A seven-transmembrane protein-TM7SF3, resides in nuclear speckles and regulates alternative splicing.
A seven-transmembrane protein-TM7SF3, resides in nuclear speckles and regulates alternative splicing.
iScience (2022-10-29)
Roi Isaac, Yaron Vinik, Martin Mikl, Shani Nadav-Eliyahu, Hadas Shatz-Azoulay, Adi Yaakobi, Natalie DeForest, Amit R Majithia, Nicholas J G Webster, Yaron Shav-Tal, Eytan Elhanany, Yehiel Zick
ABSTRACT
The seven-transmembrane superfamily member 3 protein (TM7SF3) is a p53-regulated homeostatic factor that attenuates cellular stress and the unfolded protein response. Here we show that TM7SF3 localizes to nuclear speckles; eukaryotic nuclear bodies enriched in splicing factors. This unexpected location for a trans -membranal protein enables formation of stable complexes between TM7SF3 and pre-mRNA splicing factors including DHX15, LARP7, HNRNPU, RBM14, and HNRNPK. Indeed, TM7SF3 regulates alternative splicing of >330 genes, mainly at the 3'end of introns by directly modulating the activity of splicing factors such as HNRNPK. These effects are observed both in cell lines and primary human pancreatic islets. Accordingly, silencing of TM7SF3 results in differential expression of 1465 genes (about 7% of the human genome); with 844 and 621 genes being up- or down-regulated, respectively. Our findings implicate TM7SF3, as a resident protein of nuclear speckles and suggest a role for seven-transmembrane proteins as regulators of alternative splicing.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Anti-hnRNP U Antibody, clone 3G6, clone 3G6, from mouse