Merck
  • Isthmin-1 is an adipokine that promotes glucose uptake and improves glucose tolerance and hepatic steatosis.
Isthmin-1 is an adipokine that promotes glucose uptake and improves glucose tolerance and hepatic steatosis.
Cell metabolism (2021-08-05)
Zewen Jiang, Meng Zhao, Laetitia Voilquin, Yunshin Jung, Mari A Aikio, Tanushi Sahai, Florence Y Dou, Alexander M Roche, Ivan Carcamo-Orive, Joshua W Knowles, Martin Wabitsch, Eric A Appel, Caitlin L Maikawa, Joao Paulo Camporez, Gerald I Shulman, Linus Tsai, Evan D Rosen, Christopher D Gardner, Bruce M Spiegelman, Katrin J Svensson
ABSTRACT
With the increasing prevalence of type 2 diabetes and fatty liver disease, there is still an unmet need to better treat hyperglycemia and hyperlipidemia. Here, we identify isthmin-1 (Ism1) as an adipokine and one that has a dual role in increasing adipose glucose uptake while suppressing hepatic lipid synthesis. Ism1 ablation results in impaired glucose tolerance, reduced adipose glucose uptake, and reduced insulin sensitivity, demonstrating an endogenous function for Ism1 in glucose regulation. Mechanistically, Ism1 activates a PI3K-AKT signaling pathway independently of the insulin and insulin-like growth factor receptors. Notably, while the glucoregulatory function is shared with insulin, Ism1 counteracts lipid accumulation in the liver by switching hepatocytes from a lipogenic to a protein synthesis state. Furthermore, therapeutic dosing of recombinant Ism1 improves diabetes in diet-induced obese mice and ameliorates hepatic steatosis in a diet-induced fatty liver mouse model. These findings uncover an unexpected, bioactive protein hormone that might have simultaneous therapeutic potential for diabetes and fatty liver disease.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder