Merck
  • Canine induced pluripotent stem cell maintenance under feeder-free and chemically-defined conditions.
Canine induced pluripotent stem cell maintenance under feeder-free and chemically-defined conditions.
Molecular reproduction and development (2021-05-20)
Kazuto Kimura, Masaya Tsukamoto, Takumi Yoshida, Miyuu Tanaka, Mitsuru Kuwamura, Manami Ohtaka, Ken Nishimura, Mahito Nakanishi, Kikuya Sugiura, Shingo Hatoya
ABSTRACT
Canine induced pluripotent stem cells (ciPSCs) provide a platform for regenerative veterinary medicine, disease modeling, and drug discovery. However, in the conventional method, ciPSCs are maintained using chemically-undefined media containing unknown animal components under on-murine embryonic fibroblast feeder conditions, which were reported to modify cell surface of iPSCs and increases the risk of immune rejection when the cells are transplanted into patients. Moreover, in the conventional method, ciPSCs are mechanically passaged, which requires much time and effort. Therefore, the large-scale expansion of ciPSCs is difficult, which should be resolved for using ciPSCs in clinical application and research. Here, it was shown that StemFit® AK02N and iMatrix-511 could maintain the pluripotency of ciPSCs using conventional culture method. Furthermore, it was demonstrated that the feeder-free and chemically-defined ciPSC culture systems using StemFit® AK02N and iMatrix-511 could stably maintain and allow the easy expansion of ciPSCs generated using N2B27 and StemFit® AK02N, without causing karyotype abnormalities. ciPSCs expressed several pluripotency markers and formed teratomas, including cells derived from three germ layers.
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, clone MC-480, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®