- Myc controls a distinct transcriptional program in fetal thymic epithelial cells that determines thymus growth
Myc controls a distinct transcriptional program in fetal thymic epithelial cells that determines thymus growth
Nature Communications (2019)
Jennifer E C, et al.