Merck
Drug Repurposing for the SARS-CoV-2 Papain-Like Protease.
ChemMedChem (2021-08-24)
Chia-Chuan Cho, Shuhua G Li, Tyler J Lalonde, Kai S Yang, Ge Yu, Yuchen Qiao, Shiqing Xu, Wenshe Ray Liu
ABSTRACT
As the pathogen of COVID-19, SARS-CoV-2 encodes two essential cysteine proteases that process the pathogen's two large polypeptide products pp1a and pp1ab in the human cell host to form 15 functionally important, mature nonstructural proteins. One of the two enzymes is papain-like protease or PLPro . It possesses deubiquitination and deISGylation activities that suppress host innate immune responses toward SARS-CoV-2 infection. To repurpose drugs for PLPro , we experimentally screened libraries of 33 deubiquitinase and 37 cysteine protease inhibitors on their inhibition of PLPro . Our results showed that 15 deubiquitinase and 1 cysteine protease inhibitors exhibit strong inhibition of PLPro at 200 μM. More comprehensive characterizations revealed seven inhibitors GRL0617, SJB2-043, TCID, DUB-IN-1, DUB-IN-3, PR-619, and S130 with an IC50 value below 40 μM and four inhibitors GRL0617, SJB2-043, TCID, and PR-619 with an IC50 value below 10 μM. Among four inhibitors with an IC50 value below 10 μM, SJB2-043 is the most unique in that it does not fully inhibit PLPro but has a noteworthy IC50 value of 0.56 μM. SJB2-043 likely binds to an allosteric site of PLPro to convene its inhibition effect, which needs to be further investigated. As a pilot study, the current work indicates that COVID-19 drug repurposing by targeting PLPro holds promise, but in-depth analysis of repurposed drugs is necessary to avoid omitting critical allosteric inhibitors.
Sigma-Aldrich
SJB2-043, ≥98% (HPLC)