Merck
  • Regulation of mitophagy by the NSL complex underlies genetic risk for Parkinson's disease at 16q11.2 and MAPT H1 loci.
Regulation of mitophagy by the NSL complex underlies genetic risk for Parkinson's disease at 16q11.2 and MAPT H1 loci.
Brain : a journal of neurology (2022-09-09)
Marc P M Soutar, Daniela Melandri, Benjamin O'Callaghan, Emily Annuario, Amy E Monaghan, Natalie J Welsh, Karishma D'Sa, Sebastian Guelfi, David Zhang, Alan Pittman, Daniah Trabzuni, Anouk H A Verboven, Kylie S Pan, Demis A Kia, Magda Bictash, Sonia Gandhi, Henry Houlden, Mark R Cookson, Nael Nadif Kasri, Nicholas W Wood, Andrew B Singleton, John Hardy, Paul J Whiting, Cornelis Blauwendraat, Alexander J Whitworth, Claudia Manzoni, Mina Ryten, Patrick A Lewis, Hélène Plun-Favreau
ABSTRACT
Parkinson's disease is a common incurable neurodegenerative disease. The identification of genetic variants via genome-wide association studies has considerably advanced our understanding of the Parkinson's disease genetic risk. Understanding the functional significance of the risk loci is now a critical step towards translating these genetic advances into an enhanced biological understanding of the disease. Impaired mitophagy is a key causative pathway in familial Parkinson's disease, but its relevance to idiopathic Parkinson's disease is unclear. We used a mitophagy screening assay to evaluate the functional significance of risk genes identified through genome-wide association studies. We identified two new regulators of PINK1-dependent mitophagy initiation, KAT8 and KANSL1, previously shown to modulate lysine acetylation. These findings suggest PINK1-mitophagy is a contributing factor to idiopathic Parkinson's disease. KANSL1 is located on chromosome 17q21 where the risk associated gene has long been considered to be MAPT. While our data do not exclude a possible association between the MAPT gene and Parkinson's disease, they provide strong evidence that KANSL1 plays a crucial role in the disease. Finally, these results enrich our understanding of physiological events regulating mitophagy and establish a novel pathway for drug targeting in neurodegeneration.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Anti-KANSL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-Ubiquitin (Ser65), from rabbit, purified by affinity chromatography