Merck
  • Species differences in the localization and number of CNS beta adrenergic receptors: rat versus guinea pig.
Species differences in the localization and number of CNS beta adrenergic receptors: rat versus guinea pig.
The Journal of pharmacology and experimental therapeutics (1989-06-01)
R M Booze, E A Crisostomo, J N Davis
ABSTRACT
The localization and number of beta adrenergic receptors were directly compared in the brains of rats and guinea pigs. The time course of association and saturability of [125I]cyanopindolol (CYP) binding to slide-mounted tissue sections was similar in rats (Kd = 17 pM) and guinea pigs (Kd = 20 pM). The beta-1 and beta-2 receptor subtypes were examined through the use of highly selective unlabeled receptor antagonists, ICI 118,551 (50 nM) and ICI 89,406 (70 nM). Dramatic species differences between rats and guinea pigs were observed in the neuroanatomical regional localization of the beta adrenergic receptor subtypes. For example, in the thalamus prominent beta-1 and beta-2 receptor populations were identified in the rat; however, the entire thalamus of the guinea pig had few, if any, beta adrenergic receptors of either subtype. Hippocampal area CA1 had high levels of beta-2 adrenergic receptors in both rats and guinea pigs but was accompanied by a widespread distribution of beta-2 adrenergic receptors only in rats. Quantitative autoradiographic analyses of 25 selected neuroanatomical regions 1) confirmed the qualitative differences in CNS beta adrenergic receptor localization, 2) determined that guinea pigs had significantly lower levels of beta adrenergic receptors than rats and 3) indicated a differential pattern of receptor subtypes between the two species. Knowledge of species differences in receptor patterns may be useful in designing effective experiments as well as in exploring the relationships between receptor and innervation patterns. Collectively, these data suggest caution be used in extrapolation of the relationships of neurotransmitters and receptors from studies of a single species.
Sigma-Aldrich
ICI 118,551 hydrochloride, ≥98% (HPLC), powder
Pricing and availability is not currently available.