Merck
A comprehensive analysis of precursor microRNA cleavage by human Dicer.
RNA (New York, N.Y.) (2012-09-18)
Yong Feng, Xiaoxiao Zhang, Paul Graves, Yan Zeng
ABSTRACT
Dicer cleaves double-stranded RNAs (dsRNAs) or precursor microRNAs (pre-miRNAs) to yield ≈ 22-nt RNA duplexes. The pre-miRNA structure requirement for human Dicer activity is incompletely understood. By large-scale in vitro dicing assays and mutagenesis studies, we showed that human Dicer cleaves most, although not all, of the 161 tested human pre-miRNAs efficiently. The stable association of RNAs with Dicer, as examined by gel shift assays, appears important but is not sufficient for cleavage. Human Dicer tolerates remarkable structural variation in its pre-miRNA substrates, although the dsRNA feature in the stem region and the 2-nt 3'-overhang structure in a pre-miRNA contribute to its binding and cleavage by Dicer, and a large terminal loop further enhances pre-miRNA cleavage. Dicer binding protects the terminal loop from digestion by S1 nuclease, suggesting that Dicer interacts directly with the terminal loop region.
Sigma-Aldrich
Nuclease S1 from Aspergillus oryzae, for single-strand DNA/RNA digestion
Pricing and availability is not currently available.
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)
Pricing and availability is not currently available.