Merck
  • Herbivore induction of jasmonic acid and chemical defences reduce photosynthesis in Nicotiana attenuata.
Herbivore induction of jasmonic acid and chemical defences reduce photosynthesis in Nicotiana attenuata.
Journal of experimental botany (2012-12-25)
Paul D Nabity, Jorge A Zavala, Evan H DeLucia
ABSTRACT
Herbivory initiates a shift in plant metabolism from growth to defence that may reduce fitness in the absence of further herbivory. However, the defence-induced changes in carbon assimilation that precede this reallocation in resources remain largely undetermined. This study characterized the response of photosynthesis to herbivore induction of jasmonic acid (JA)-related defences in Nicotiana attenuata to increase understanding of these mechanisms. It was hypothesized that JA-induced defences would immediately reduce the component processes of photosynthesis upon attack and was predicted that wild-type plants would suffer greater reductions in photosynthesis than plants lacking JA-induced defences. Gas exchange, chlorophyll fluorescence, and thermal spatial patterns were measured together with the production of defence-related metabolites after attack and through recovery. Herbivore damage immediately reduced electron transport and gas exchange in wild-type plants, and gas exchange remained suppressed for several days after attack. The sustained reductions in gas exchange occurred concurrently with increased defence metabolites in wild-type plants, whereas plants lacking JA-induced defences suffered minimal suppression in photosynthesis and no increase in defence metabolite production. This suppression in photosynthesis occurred only after sustained defence signalling and defence chemical mobilization, whereas a short bout of feeding damage only transiently altered components of photosynthesis. It was identified that lipoxygenase signalling interacted with photosynthetic electron transport and that the resulting JA-related metabolites reduced photosynthesis. These data represent a metabolic cost to mounting a chemical defence against herbivory and link defence-signalling networks to the differential effects of herbivory on photosynthesis in remaining leaf tissues in a time-dependent manner.
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type I-B, lyophilized powder, ≥50,000 units/mg solid