Merck
  • Acute aortocaval fistula: role of low perfusion pressure and subendocardial remodeling on left ventricular function.
Acute aortocaval fistula: role of low perfusion pressure and subendocardial remodeling on left ventricular function.
International journal of experimental pathology (2013-04-19)
Flávia R R Mazzo, Clovis de Carvalho Frimm, Ana Iochabel S Moretti, Maria C Guido, Marcia K Koike
ABSTRACT
The experimental model of aortocaval fistula is a useful model of cardiac hypertrophy in response to volume overload. In the present study it has been used to investigate the pathologic subendocardial remodeling associated with the development of heart failure during the early phases (day 1, 3, and 7) following volume overload. Compared with sham treated rats, aortocaval fistula rats showed lower systemic blood pressure and higher left ventricular end-diastolic pressure This resulted in lower coronary driving pressure and left ventricular systolic and diastolic dysfunction. Signs of myocyte necrosis, leukocyte cell infiltration, fibroplasia and collagen deposition appeared sequentially in the subendocardium where remodeling was more prominent than in the non-subendocardium. Accordingly, increased levels of TNF-alpha, IL-1 beta, and IL-6, and enhanced MMP-2 activity were all found in the subendocardium of rats with coronary driving pressure ≤ 60 mmHg. The coronary driving pressure was inversely correlated with MMP-2 activity in subendocardium in all time-points studied, and blood flow in this region showed positive correlation with systolic and diastolic function at day 7. Thus the predominant subendocardial remodeling that occurs in response to low myocardial perfusion pressure during the acute phases of aortocaval fistula contributes to early left ventricular dysfunction.
Sigma-Aldrich
Peroxidase from horseradish, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Lactoperoxidase from bovine milk, lyophilized, powder, ≥150 U/mg
Sigma-Aldrich
Peroxidase from horseradish, Type X, ammonium sulfate suspension
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Lactoperoxidase from bovine milk, lyophilized powder (essentially salt-free), ≥200 units/mg protein
Sigma-Aldrich
Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Myeloperoxidase from human leukocytes, lyophilized powder, ≥50 units/mg protein