Merck
  • Simultaneous determination of swertiamarin and its metabolites (5Z)-5-ethylidene-8-hydroxy-3,4,5,6,7,8-hexahydro-1H-pyrano[3,4-c]pyridin-1-one and erythrocentaurin in broth of Aspergillus niger by HPLC.
Simultaneous determination of swertiamarin and its metabolites (5Z)-5-ethylidene-8-hydroxy-3,4,5,6,7,8-hexahydro-1H-pyrano[3,4-c]pyridin-1-one and erythrocentaurin in broth of Aspergillus niger by HPLC.
Biomedical chromatography : BMC (2007-09-14)
Chang Jun, Zhao Xue-Ming, Liu Chang-Xiao, Zhang Tie-Jun
ABSTRACT
When cultivated with Aspergillus niger, swertiamarin, an important drug, is rapidly transformed into erythrocentaurin and (5Z)-5-ethylidene-8-hydroxy-3,4,5,6,7,8-hexahydro-1H-pyrano[3,4-c]pyridin-1-one (M(1)), a new compound with high anti-inflammatory activity. A simple and rapid HPLC method for simultaneous determination of swertiamarin and its two metabolites in broth of A. niger is described. The chromatographic separation was achieved on a C(18) ODS column (250 x 4.6 mm i.d.) by gradient elution with 0.04% formic acid in water and 0.04% formic acid in acetonitrile as the gradient mixtures. The flow rate was 1 mL/min, the detection wavelength was 237 nm and the column temperature was kept at 30 degrees C. The retention times of swertiamarin, erythrocentaurin and M(1) were 14.6, 16.8 and 24.8 min, respectively. The mean absolute recoveries of three analysts were over 96%. Quantification limits were 0.02 microg/mL for swertiamarin and 0.05 microg/mL for both of the two metabolites. The method was applied for the quantification of swertiamarin and its two metabolites during the fermentation process and the evaluation of the bioavailabilities in the Caco-2 monolayer.
Sigma-Aldrich
Swertiamarin, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.