Merck
Antihyperlipidemic compounds from the fruit of Piper longum L.
Phytotherapy research : PTR (2009-01-28)
Zhuang Jin, Gereltu Borjihan, Ruiguo Zhao, Zhaorigetu Sun, Gerald B Hammond, Toshiyuki Uryu
ABSTRACT
A bioassay-guided isolation of an ethanol extract of the fruit of Piper longum L. yielded piperlonguminine, piperine and pipernonaline, as the main antihyperlipidemic constituents. They exhibit appreciable antihyperlipidemic activity in vivo, which is comparable to that of the commercial antihyperlipidemic drug, simvastatin.
Sigma-Aldrich
Pipernonaline, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.