Merck
  • Dynamical dimer structure and liquid structure of fatty acids in their binary liquid mixture: dodecanoic and 3-phenylpropionic acids system.
Dynamical dimer structure and liquid structure of fatty acids in their binary liquid mixture: dodecanoic and 3-phenylpropionic acids system.
Chemistry and physics of lipids (2004-04-15)
Makio Iwahashi, Shintaro Takebayashi, Atsushi Umehara, Yasutoshi Kasahara, Hideyuki Minami, Hideyo Matsuzawa, Tohru Inoue, Hiroshi Takahashi
ABSTRACT
Dimer structure and liquid structure of fatty acids in the binary liquid mixture of dodecanoic (LA) and 3-phenylpropionic acids (PPA) were studied through the measurements of DSC, self-diffusion coefficient (D), density, viscosity, 13C NMR spin-lattice relaxation time, small-angle X-ray scattering (SAXS), and small-angle neutron scattering (SANS). The phase diagram of LA/PPA mixture exhibited a typical eutectic pattern, which means that LA and PPA are completely immiscible in solid phase. In the liquid phase of the LA/PPA mixture, D of LA always differed from that of PPA irrespective of their compositions. This exhibited that, in the liquid phase of the binary mixture of fatty acids giving a complete eutectic in the solid phase, the fatty acid dimers are composed of the same fatty acid species irrespective of their compositions. The liquid structure of the LA/PPA mixture was clarified through the SAXS and also the SANS measurements.
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG