Merck
  • Frontocortical 5-HT4 receptors exert positive feedback on serotonergic activity: viral transfections, subacute and chronic treatments with 5-HT4 agonists.
Frontocortical 5-HT4 receptors exert positive feedback on serotonergic activity: viral transfections, subacute and chronic treatments with 5-HT4 agonists.
Biological psychiatry (2005-04-12)
Guillaume Lucas, Valérie Compan, Yves Charnay, Rachael L Neve, Eric J Nestler, Joël Bockaert, Michel Barrot, Guy Debonnel
ABSTRACT
We recently identified a facilitory control exerted by serotonin4 (5-HT4) receptors on the in vivo firing activity of dorsal raphe nucleus (DRN) serotonergic (5-HT) neurons. However, these findings were based on acute administrations of 5-HT4 receptor agonists and antagonists, which were active only in a subpopulation of 5-HT neurons. We had no evidence that this influence was significant when considering the entire DRN, nor if it was persistent after chronic treatments. In addition, the poor distribution of 5-HT4 receptors within the DRN raised the question of the neuroanatomical bases underlying this control. Here we show that the subacute intraperitoneal (IP) injection of the 5-HT4 receptor agonists prucalopride (2.5 mg/kg) and RS 67333 (1.5 mg/kg) 30 minutes before the beginning of recordings augment the mean firing rate of DRN neurons by 40% and 66%, respectively. These increases remain stable when the compounds are administered continuously during 3 and 21 days; the effects of the 3-day treatment are blocked by the 5-HT4 receptor antagonist GR 125487 (1000 microg/kg, intravenous [i.v.]). In addition, stereotaxic microinjections of herpes simplex viruses, transformed to overexpress 5-HT4 receptors, increase DRN 5-HT neuronal mean activity when performed in the medial prefrontal cortex (mPFC) but not in the striatum or in the hippocampus. This finding suggests the existence of a 5-HT(4)-dependent activation of DRN that may involve the mPFC, unveiling the 5-HT4 receptor as a putative player in the physiopathology of several disorders related to central 5-HT dysfunction.
Sigma-Aldrich
GR 113808, ≥98% (HPLC), solid
Pricing and availability is not currently available.