Merck
  • Mutagenic, antimutagenic and antioxidant activities of a new class of beta-glucoside hydroxyhydroquinone from Anagallis monelli growing in Tunisia.
Mutagenic, antimutagenic and antioxidant activities of a new class of beta-glucoside hydroxyhydroquinone from Anagallis monelli growing in Tunisia.
Chemical & pharmaceutical bulletin (2007-03-03)
Samia Ammar, Mohamed Ali Mahjoub, Nesrine Charfi, Ines Skandarani, Leila Chekir-Ghedira, Zine Mighri
ABSTRACT
A new skeleton of an O-heteroside natural substance named zinolol, the first representative of a new class of aminated hydroxyhydroquinone, has been isolated from the whole plant Anagallis monelli. Its structure has been established by one and two dimensional NMR spectroscopic procedures. Antioxidant, mutagenic, antimutagenic activities were realised and positive results were recorded.
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%