Merck
  • Rapid determination of ETS markers with a prototype field-portable GC employing a microsensor array detector.
Rapid determination of ETS markers with a prototype field-portable GC employing a microsensor array detector.
Journal of environmental monitoring : JEM (2007-05-12)
Qiongyan Zhong, Rebecca A Veeneman, William H Steinecker, Chunrong Jia, Stuart A Batterman, Edward T Zellers
ABSTRACT
The adaptation of a portable gas chromatograph (GC) prototype with several unique design features to the determination of vapor-phase markers of environmental tobacco smoke (ETS) is described. This instrument employs a dual-stage adsorbent preconcentrator, two series-coupled separation columns that can be independently temperature programmed, and a detector consisting of an array of nanoparticle-coated chemiresistors, whose response patterns are used together with retention times for vapor recognition. An adsorbent pre-trap was developed to remove semi-volatile organics from the sample stream. Conditions were established to quantitatively capture two ETS markers, 2,5-dimethylfuran (2,5-DMF) and 4-ethenylpyridine (4-EP, as a surrogate for 3-EP), and to separate them from the 34 most prominent co-contaminants present in ETS using ambient air as the carrier gas. A complete analysis can be performed every 15 min. Projected detection limits are 0.58 and 0.08 ppb for 2,5-DMF and 4-EP, respectively, assuming a 1 L sample volume, which are sufficiently low to determine these markers in typical smoking-permitted environments.
Sigma-Aldrich
2,5-Dimethylfuran, 99%