Merck
  • Cloning and characterization of a new laccase from Bacillus licheniformis catalyzing dimerization of phenolic acids.
Cloning and characterization of a new laccase from Bacillus licheniformis catalyzing dimerization of phenolic acids.
Applied microbiology and biotechnology (2008-03-12)
Katja Koschorreck, Sven M Richter, Augusta B Ene, Emil Roduner, Rolf D Schmid, Vlada B Urlacher
ABSTRACT
A new laccase gene (cotA) was cloned from Bacillus licheniformis and expressed in Escherichia coli. The recombinant protein CotA was purified and showed spectroscopic properties, typical for blue multi-copper oxidases. The enzyme has a molecular weight of approximately 65 kDa and demonstrates activity towards canonical laccase substrates 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) (ABTS), syringaldazine (SGZ) and 2,6-dimethoxyphenol (2,6-DMP). Kinetic constants KM and kcat for ABTS were of 6.5+/-0.2 microM and 83 s(-1), for SGZ of 4.3+/-0.2 microM and 100 s(-1), and for 2,6-DMP of 56.7+/-1.0 microM and 28 s(-1). Highest oxidizing activity towards ABTS was obtained at 85 degrees C. However, after 1 h incubation of CotA at 70 degrees C and 80 degrees C, a residual activity of 43% and 8%, respectively, was measured. Furthermore, oxidation of several phenolic acids and one non-phenolic acid by CotA was investigated. CotA failed to oxidize coumaric acid, cinnamic acid, and vanillic acid, while syringic acid was oxidized to 2,6-dimethoxy-1,4-benzoquinone. Additionally, dimerization of sinapic acid, caffeic acid, and ferulic acid by CotA was observed, and highest activity of CotA was found towards sinapic acid.
Sigma-Aldrich
Syringaldazine, indicator for laccase and peroxidase activity
Sigma-Aldrich
Syringaldazine, 98%