Merck
  • Synthesis and structure-activity relationships of long-acting beta2 adrenergic receptor agonists incorporating arylsulfonamide groups.
Synthesis and structure-activity relationships of long-acting beta2 adrenergic receptor agonists incorporating arylsulfonamide groups.
Journal of medicinal chemistry (2009-03-26)
Panayiotis A Procopiou, Victoria J Barrett, Nicola J Bevan, Keith Biggadike, Peter R Butchers, Diane M Coe, Richard Conroy, Dean D Edney, Rita N Field, Alison J Ford, Stephen B Guntrip, Brian E Looker, Iain M McLay, Michael J Monteith, Valerie S Morrison, Peter J Mutch, Stephen A Richards, Rosemary Sasse, Claire E Smith
ABSTRACT
A series of saligenin alkoxyalkylphenylsulfonamide beta(2) adrenoceptor agonists were prepared by reacting a protected saligenin oxazolidinone with alkynyloxyalkyl bromides, followed by Sonogashira reaction, hydrogenation, and deprotection. The meta-substituted primary sulfonamide was more potent than the para- and the ortho-analogues. Primary sulfonamides were more potent than the secondary and tertiary analogues. The onset and duration of action in vitro of selected compounds was assessed on isolated superfused guinea pig trachea. Sulfonamide 29b had the best profile of potency, selectivity, onset, and duration of action on both guinea pig trachea and human bronchus. Furthermore, 29b was found to have low oral bioavailability in rat and dog and also to have long duration of action in an in vivo model of bronchodilation. Crystalline salts of 29b were identified that had suitable properties for inhaled administration. A proposed binding mode for 29b to the beta(2)-receptor is presented.
Sigma-Aldrich
2-Amino-1-phenylethanol, 98%
Pricing and availability is not currently available.