Merck
  • Rapid and complete degradation of the herbicide picloram by Lipomyces kononenkoae.
Rapid and complete degradation of the herbicide picloram by Lipomyces kononenkoae.
Journal of agricultural and food chemistry (2009-06-06)
Michael J Sadowsky, William C Koskinen, Marianne Bischoff, Brian L Barber, Joanna M Becker, Ronald F Turco
ABSTRACT
An enrichment culture approach was used to isolate a pure culture of the yeast Lipomyces kononenkoae, which had the ability to grow on the herbicide picloram. The yeast rapidly and completely degraded 50 microg mL(-1) picloram by 48 h of growth. While L. kononenkoae was found to use both N atoms of picloram as a sole nitrogen source for growth, it failed to mineralize the herbicide or use it as a sole C source. Product analysis done using LC-ESI-MS indicated that biodegradation of picloram by L. kononenkoae proceeds via a didechlorinated, dihydroxylated, pyridinecarboxylic acid derivative. Our results are consistent with the hypothesis that the majority of picloram degradation in the soil is likely due to microbial catabolic processes.
Product Description
Supelco
Picloram, PESTANAL®, analytical standard
Sigma-Aldrich
Picloram, BioReagent, suitable for plant cell culture