Merck
  • Premature pubarche in Mediterranean girls: high prevalence of heterozygous CYP21 mutation carriers.
Premature pubarche in Mediterranean girls: high prevalence of heterozygous CYP21 mutation carriers.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology (2010-01-12)
Françoise Paris, Véronique Tardy, Anne Chalançon, Marie Christine Picot, Yves Morel, Charles Sultan
ABSTRACT
This study investigated the prevalence and consequences of heterozygous CYP21A2 mutations in premature pubarche (PP) girls. We investigated 36 French Mediterranean girls with isolated PP. We performed synacthen testing with 17OHP and 21-deoxycortisol evaluation, along with molecular analysis of the CYP21A2 gene in girls with abnormal elevation of one of these two adrenal steroids. Three girls (8.3%) had nonclassical adrenal hyperplasia, secondary to compound heterozygosity that associated at least one severe mutation for the three girls. A heterozygous mutation of the CYP21A2 gene was confirmed by molecular biology in eight girls (22%); a deletion of the CYP21A2 gene was found in one of them. Biological hyperandrogenism was found in the prepubertal CYP21A2 mutation carriers, whereas the four heterozygous girls who were followed long enough to have reached pubertal age presented biological and clinical hyperandrogenism. We underline the high prevalence of heterozygous CYP21A2 mutations in girls with PP and demonstrate the usefulness of systematic screening by synacthen testing, both to improve their future clinical management and to prevent the transmission of classical adrenal hyperplasia to future offspring. Because of the severe metabolic and cardiovascular consequences of hyperandrogenism, long-term follow-up of these heterozygous patients is mandatory.
Sigma-Aldrich
Reichstein′s substance S, ≥98%
Supelco
21-Deoxycortisol solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
21-Deoxycortisol
Supelco
11-Deoxycortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®