Merck
  • Simultaneous production of p-tolualdehyde and hydrogen peroxide in photocatalytic oxygenation of p-xylene and reduction of oxygen with 9-mesityl-10-methylacridinium ion derivatives.
Simultaneous production of p-tolualdehyde and hydrogen peroxide in photocatalytic oxygenation of p-xylene and reduction of oxygen with 9-mesityl-10-methylacridinium ion derivatives.
Chemical communications (Cambridge, England) (2010-01-12)
Kei Ohkubo, Kentaro Mizushima, Ryosuke Iwata, Kazunori Souma, Nobuo Suzuki, Shunichi Fukuzumi
ABSTRACT
Photooxygenation of p-xylene by oxygen occurs efficiently under photoirradiation of 9-mesityl-2,7,10-trimethylacridinium ion (Me(2)Acr(+)-Mes) to yield p-tolualdehyde and hydrogen peroxide, which is initiated via photoinduced electron transfer of Me(2)Acr(+)-Mes to produce the electron-transfer state.
Sigma-Aldrich
p-Tolualdehyde, ≥97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
p-Tolualdehyde, 97%
Pricing and availability is not currently available.