Merck
  • Preparation and orcein staining of mitotic chromosomes from Drosophila larval brain.
Preparation and orcein staining of mitotic chromosomes from Drosophila larval brain.
Cold Spring Harbor protocols (2010-03-03)
Sergio Pimpinelli, Silvia Bonaccorsi, Laura Fanti, Maurizio Gatti
ABSTRACT
In this protocol, larval brains from Drosophila are incubated in vitro with colchicine, treated with hypotonic solution, fixed, and squashed in aceto-orcein. This procedure provides a large number of well-spread metaphase figures (200-400 per brain) that can be analyzed for chromosome morphology, the presence of chromosome aberrations, and the degree of ploidy.
Sigma-Aldrich
Orcein, synthetic
Sigma-Aldrich
Orcein, synthetic, certified by the BSC