Merck
  • The performance of 1-ethyl-3-methylimidazolium tetrafluoroborate ionic liquid as mobile phase additive in HPLC-based lipophilicity assessment.
The performance of 1-ethyl-3-methylimidazolium tetrafluoroborate ionic liquid as mobile phase additive in HPLC-based lipophilicity assessment.
Biomedical chromatography : BMC (2010-08-27)
Costas Giaginis, Anna Tsantili-Kakoulidou
ABSTRACT
Ionic liquids have been widely used as green alternative mobile phase additives to shield the residuals silanols groups and modify the stationary/mobile phase HPLC systems. The present study aimed to evaluate the performance of the ionic liquid 1-ethyl-3-methylimidazolium tetrafluoroborate ([EMIM][BF4]) in producing extrapolated logk(w) indices suitable to substitute for octanol-water logP or logD values. The effect of [EMIM][BF4] was investigated for a set of basic and neutral drugs using two different columns, BDS and ABZ(+) . [EMIM][BF4] was added simply alone or in combination with n-octanol and was compared with the conventional masking agent n-decylamine. [EMIM][BF4] reduced the retention by suppressing silanophilic interactions, althoug to a lower extent than n-decylamine. Addition of n-octanol further decreased the retention by shielding silanol sites on BDS and/or interacting with polar groups through hydrogen bonding on ABZ(+) . Logk(w) /logD(7.4) relationships proved moderate compared with those derived upon addition of n-decylamine. They were considerably improved upon the introduction of protonated fraction F(+) in the correlation, reflecting ion pair formation between the chaotropic anion [BF4](-) and the protonated basic compounds. In this aspect, the ionic liquid [EMIM][BF4], although efficient as a masking agent, cannot be recommended as mobile phase additive to reproduce octanol-water partitioning.
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Decylamine, ≥99.0% (GC)