Merck
  • Synthesis and in vitro activity of N-benzyl-1-(2,3-dichlorophenyl)-1H-tetrazol-5-amine P2X(7) antagonists.
Synthesis and in vitro activity of N-benzyl-1-(2,3-dichlorophenyl)-1H-tetrazol-5-amine P2X(7) antagonists.
Bioorganic & medicinal chemistry letters (2011-05-04)
Arturo Perez-Medrano, Diana L Donnelly-Roberts, Alan S Florjancic, Derek W Nelson, Tongmei Li, Marian T Namovic, Sridhar Peddi, Connie R Faltynek, Michael F Jarvis, William A Carroll
ABSTRACT
Synthesis and biological evaluation of a novel class of substituted N-benzyl-1-(2,3-dichlorophenyl)-1H-tetrazol-5-amine derivatives resulted in the identification of potent P2X(7) antagonists. These compounds were assayed for activity at both the human and rat P2X(7) receptors. On the benzyl moiety, a variety of functional groups were tolerated, including both electron-withdrawing and electron-donating substituents. Ortho-substitution on the benzyl group provided the greatest potency. The ortho-substituted analogs showed approximately 2.5-fold greater potency at human compared to rat P2X(7) receptors. Compounds 12 and 38 displayed hP2X(7)pIC(50)s>7.8 with less than 2-fold difference in potency at the rP2X(7).
Sigma-Aldrich
Tetrazole solution, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile