Merck
  • Stepwise preparation and characterization of molecular wires made of zinc octaethylporphyrin complexes bridged by 4,4'-bipyridine on HOPG.
Stepwise preparation and characterization of molecular wires made of zinc octaethylporphyrin complexes bridged by 4,4'-bipyridine on HOPG.
Nanotechnology (2011-10-06)
Quirina Ferreira, Luís Alcácer, Jorge Morgado
ABSTRACT
Molecular-scale devices can be made using a step-by-step procedure, in a controllable and highly versatile way. In this report, we describe the growth of molecular wires (MW) from zinc (II) octaethylporphyrin (ZnOEP) assembled on highly oriented pyrolytic graphite (HOPG) by a step-by-step approach using 4,4'-bipyridine (BP) to bridge the porphyrin units, via coordination of the nitrogen atom to zinc. In order to gain an insight into the molecular self-organization of these wires, we carried out a detailed scanning tunnelling microscopy (STM) analysis of each monolayer, using a solid/liquid interface technique, up to a complete ZnOEP/BP/ZnOEP/BP/ZnOEP-assembled structure. The electrical properties of the MWs were assessed by scanning tunnelling spectroscopy (STS) and by current-sensing atomic force microscopy (CS-AFM), showing an increase of electrical resistance with the length of the MW.
Sigma-Aldrich
4,4′−Dipyridyl, 98%