Merck
  • Composition and antibacterial activity of essential oil from Scutellaria grossa Wall ex Benth.
Composition and antibacterial activity of essential oil from Scutellaria grossa Wall ex Benth.
Natural product research (2011-11-09)
Charu C Pant, Anand B Melkani, Lalit Mohan, Vasu Dev
ABSTRACT
The chemical composition of the steam volatile oil of the whole aerial part of Scutellaria grossa Wall ex Benth. (Lamiaceae), obtained by steam distillation, has been analysed by capillary GC and GC-MS. The oil was found to be rich in oxygenated monoterpenes (88.6%). A total of 50 constituents were identified, representing 94.4% of the total oil composition. Linalool (37.0%) and 1-octen-3-ol (32.0%) were found to be the principal constituents. The antibacterial activity of the oil was determined against 10 bacterial strains by measuring the growth inhibitory zones. The oil showed significant antibacterial activity against Gram-positive bacteria, Bacillus subtilis and Enterococcus faecalis, and the Gram-negative, Klebsiella pneumoniae and Salmonella enterica enterica. A minimum inhibitory concentration of 31.25 µL mL(-1) was observed against E. faecalis.
Sigma-Aldrich
1-Octen-3-ol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octen-3-ol, 98%