Merck
  • Symmetry distortion in the human hemoglobin tetramer induced by asymmetric ligation.
Symmetry distortion in the human hemoglobin tetramer induced by asymmetric ligation.
FEBS letters (2011-12-07)
Naoya Shibayama
ABSTRACT
To investigate the conformational changes in human tetrameric (αβ)(2) hemoglobin upon binding of the first two ligands, we have measured the kinetics of reactions between 4,4'-dithiodipyridine and β93Cys sulfhydryl groups of four diliganded hemoglobins by using CO-bound Fe(II)-Ni(II) hybrids with and without β-β cross-linking. The data show that all the diliganded intermediates have high sulfhydryl reactivities, which are greater than or equal to that for the fully-liganded end state, especially when containing liganded α subunit(s). The results also reveal that both the asymmetrically (α1β1 and α1β2) diliganded species show similar high rates of sulfhydryl reactivity and biphasic kinetics, suggesting a new conformation but only slight functional distortion caused by asymmetric ligation.
Sigma-Aldrich
Aldrithiol-4, 98%