Merck
  • Once-daily application of calcipotriene 0.005%-betamethasone dipropionate 0.064% ointment for repigmentation of facial vitiligo.
Once-daily application of calcipotriene 0.005%-betamethasone dipropionate 0.064% ointment for repigmentation of facial vitiligo.
Cutis (2012-01-26)
Marissa D Newman, Nanette B Silverberg
ABSTRACT
Vitiligo vulgaris is an autoimmune pigmentary disorder with no universally efficacious therapeutic options. Separate applications of calcipotriene ointment 0.005% and topical corticosteroid ointments have been successful in the repigmentation of vitiligo. We sought to examine the efficacy of a combination calcipotriene 0.005%-betamethasone dipropionate 0.064% ointment in the repigmentation of vitiligo. An institutional review board-approved retrospective chart review was conducted in 13 pediatric and adult patients with vitiligo treated with calcipotriene 0.005%-betamethasone dipropionate 0.064% ointment once daily for at least 2 months. Two of 3 children had 76% to 100% repigmentation of facial vitiligo with once-daily usage after 2 months. Of the 10 adults (aged 28-55 years), 1 had 100% facial repigmentation in 3 months, 1 had 76% to 99% facial repigmentation in 5 to 9 months, and 2 had 26% to 50% repigmentation in 3 months. Twelve patients developed some facial repigmentation. No patients experienced atrophy, telangiectases, or lesion enlargement during treatment. Combination calcipotriene 0.005%-betamethasone dipropionate 0.064% ointment shows promise as a once-daily vitiligo therapy. Adult and pediatric facial vitiligo patients may see repigmentation as early as 2 months after initiation of therapy. Children may experience a better response, but larger studies are needed.
Product Description
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betamethasone 17,21-dipropionate