Merck
  • Highly selective carbon dioxide uptake by [Cu(bpy-n)2(SiF6)] (bpy-1 = 4,4'-bipyridine; bpy-2 = 1,2-bis(4-pyridyl)ethene).
Highly selective carbon dioxide uptake by [Cu(bpy-n)2(SiF6)] (bpy-1 = 4,4'-bipyridine; bpy-2 = 1,2-bis(4-pyridyl)ethene).
Journal of the American Chemical Society (2012-02-10)
Stephen D Burd, Shengqian Ma, Jason A Perman, Benjamin J Sikora, Randall Q Snurr, Praveen K Thallapally, Jian Tian, Lukasz Wojtas, Michael J Zaworotko
ABSTRACT
A previously known class of porous coordination polymer (PCP) of formula [Cu(bpy-n)(2)(SiF(6))] (bpy-1 = 4,4'-bipyridine; bpy-2 = 1,2-bis(4-pyridyl)ethene) has been studied to assess its selectivity toward CO(2), CH(4), N(2), and H(2)O. Gas sorption measurements reveal that [Cu(bpy-1)(2)(SiF(6))] exhibits the highest uptake for CO(2) yet seen at 298 K and 1 atm by a PCP that does not contain open metal sites. Significantly, [Cu(bpy-1)(2)(SiF(6))] does not exhibit particularly high uptake under the same conditions for CH(4), N(2), and, H(2)O, presumably because of its lack of open metal sites. Consequently, at 298 K and 1 atm [Cu(bpy-1)(2)(SiF(6))] exhibits a relative uptake of CO(2) over CH(4) of ca. 10.5:1, the highest value experimentally observed in a compound without open metal sites. [Cu(bpy-2)(2)(SiF(6))] exhibits larger pores and surface area than [Cu(bpy-1)(2)(SiF(6))] but retains a high CO(2)/CH(4) relative uptake of ca. 8:1.
Sigma-Aldrich
4,4′−Dipyridyl, 98%