Merck
  • Amine-reactive polymers synthesized by RAFT polymerization using an azlactone functional trithiocarbonate RAFT agent.
Amine-reactive polymers synthesized by RAFT polymerization using an azlactone functional trithiocarbonate RAFT agent.
Macromolecular rapid communications (2012-07-13)
Hien The Ho, Flavien Leroux, Sagrario Pascual, Véronique Montembault, Laurent Fontaine
ABSTRACT
A new azlactone-derived trithiocarbonate is prepared and used as a chain-transfer agent to mediate the reversible addition-fragmentation chain transfer (RAFT) polymerization of styrene, ethyl acrylate, and N-isopropyl acrylamide. Well-defined polymers with controlled molecular weights (M(n) = 1000-7000 g mol(-1) ) and narrow molecular weight distributions (PDI = 1.05-1.10) are thus obtained that retain the azlactone functionality at the chain end. The ability of the resulting end-functionalized polymers to react quantitatively at room temperature with a stoichiometric amount of amino groups with retention of the thiocarbonylthio moiety is ascertained by using 4-fluorobenzylamine and allylamine.
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized