Merck
  • Diastereoselective intermolecular ene reactions: synthesis of 4,5,6,7-tetrahydro-1H-benzo[d]imidazoles.
Diastereoselective intermolecular ene reactions: synthesis of 4,5,6,7-tetrahydro-1H-benzo[d]imidazoles.
Organic & biomolecular chemistry (2012-07-18)
Lynsey J Watson, Ross W Harrington, William Clegg, Michael J Hall
ABSTRACT
The Diels-Alder cycloadducts of 4-vinylimidazoles and N-phenylmaleimide are shown to undergo facile intermolecular ene reactions. Overall the reaction of three simple molecules (a diene, a dienophile and an enophile) in a two-step process gives 4,5,6,7-tetrahydro-1H-benzo[d]imidazoles with high yields, high atom economy and diastereocontrol of up to 5 new stereocentres.
Sigma-Aldrich
1-Vinylimidazole, ≥99%
Sigma-Aldrich
N-Phenylmaleimide, 97%