Merck
  • Easy introduction of maleimides at different positions of oligonucleotide chains for conjugation purposes.
Easy introduction of maleimides at different positions of oligonucleotide chains for conjugation purposes.
Organic & biomolecular chemistry (2012-09-26)
Albert Sánchez, Enrique Pedroso, Anna Grandas
ABSTRACT
[2,5-Dimethylfuran]-protected maleimides were placed at both internal positions and the 3'-end of oligonucleotides making use of solid-phase synthesis procedures. A new phosphoramidite derivative and a new solid support incorporating the protected maleimide moiety were prepared for this purpose. In all cases maleimide deprotection (retro-Diels-Alder reaction) followed by reaction with thiol-containing compounds afforded the target conjugate.
Sigma-Aldrich
2,5-Dimethylfuran, 99%