Merck
  • Giant photoluminescence enhancement in SiC nanocrystals by resonant semiconductor exciton-metal surface plasmon coupling.
Giant photoluminescence enhancement in SiC nanocrystals by resonant semiconductor exciton-metal surface plasmon coupling.
Nanotechnology (2012-12-15)
Dejian Dai, Zhenggao Dong, Jiyang Fan
ABSTRACT
We report giant fluorescence enhancement in SiC nanocrystals (NCs) embedded in a sodium dodecyl sulfonate dielectric medium by proximately contacted Ag nanoparticles. The enhancement in integrated fluorescence intensity reaches an astonishing 176-fold under 360 nm excitation (53.3-fold enhancement in emission maximum intensity). Finite-element simulation indicates that the strong resonant coupling between the excited SiC NCs and localized surface plasmons of the Ag nanoparticles plays a dominant role in determining fluorescence enhancement. In contrast, the absorption enhancement caused by light concentration around the Ag nanoparticles makes only a slight contribution to the overall enhancement. Our result opens the possibility of applications of these highly enhanced fluorescent SiC NCs in diverse areas such as sensing, optoelectronics and life sciences.
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Silicon carbide, -200 mesh particle size