Merck
  • Metabotropic NMDA receptor function is required for β-amyloid-induced synaptic depression.
Metabotropic NMDA receptor function is required for β-amyloid-induced synaptic depression.
Proceedings of the National Academy of Sciences of the United States of America (2013-02-23)
Helmut W Kessels, Sadegh Nabavi, Roberto Malinow
ABSTRACT
The mechanisms by which β-amyloid (Aβ), a peptide fragment believed to contribute to Alzheimer's disease, leads to synaptic deficits are not known. Here we find that elevated oligomeric Aβ requires ion flux-independent function of NMDA receptors (NMDARs) to produce synaptic depression. Aβ activates this metabotropic NMDAR function on GluN2B-containing NMDARs but not on those containing GluN2A. Furthermore, oligomeric Aβ leads to a selective loss of synaptic GluN2B responses, effecting a switch in subunit composition from GluN2B to GluN2A, a process normally observed during development. Our results suggest that conformational changes of the NMDAR, and not ion flow through its channel, are required for Aβ to produce synaptic depression and a switch in NMDAR composition. This Aβ-induced signaling mediated by alterations in GluN2B conformation may be a target for therapeutic intervention of Alzheimer's disease.
Sigma-Aldrich
(+)-MK-801 hydrogen maleate, powder