Merck
Toxoplasma gondii binds sheep prolactin.
Experimental parasitology (2013-03-19)
K Dzitko, B Dziadek, J Gatkowska, H Długońska
ABSTRACT
Taking into account the literature reports on the involvement of prolactin (PRL) in the regulation of immunity against Toxoplasma gondii, we decided to check whether this parasite has the ability to bind the lactotrophic hormone. We examined T. gondii binding of sheep fluoresceine- and biotine-labeled prolactin isolated from pituitary (shPRL). In this work we announced for the first time that shPRL was bound to live tachyzoites of RH (type I) and ME49 (type II) strains. Furthermore, by use of competitive inhibition analysis, we confirmed that this binding was specific for both tested T. gondii strains.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, meets USP testing specifications
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material