Merck
  • Formation of N-nitrosodiphenylamine from 1,1-diphenylhydrazine by rat liver microsomal preparations.
Formation of N-nitrosodiphenylamine from 1,1-diphenylhydrazine by rat liver microsomal preparations.
Biochemical and biophysical research communications (1984-02-14)
K Tatsumi, S Kitamura, M Sumida
ABSTRACT
The present study provides the first evidence for in vitro metabolic conversion of a 1,1-disubstituted hydrazine to the corresponding nitrosamine. The study shows that superoxide radical which is generated by NADPH-cytochrome c reductase is involved in the oxidation of 1,1-diphenylhydrazine to N-nitrosodiphenylamine catalyzed by rat liver microsomes.
Sigma-Aldrich
N,N-Diphenylhydrazine hydrochloride, 97%
Pricing and availability is not currently available.