- Teratologic evaluation of 3,6-Dichloropicolinic acid in rats and rabbits.
Teratologic evaluation of 3,6-Dichloropicolinic acid in rats and rabbits.
Fundamental and applied toxicology : official journal of the Society of Toxicology (1984-02-01)
W C Hayes, F A Smith, J A John, K S Rao
PMID6693005
ABSTRACT
3,6-Dichloropicolinic acid (clopyralid), a new herbicide, was evaluated for teratogenic potential in Fischer 344 rats and New Zealand White rabbits. Rats were given 0, 15, 75, or 250 mg clopyralid/kg/day by gavage on Days 6-15 of gestation while rabbits were given 0, 110, or 250 mg clopyralid/kg/day on Days 6-18 of gestation. Maternal toxicity, as evidenced by decreased body weight gain, was observed among pregnant rats in the 250-mg/kg/day group. No evidence of maternal toxicity was observed among treated rabbits. A teratogenic effect was not detected in either species.