Merck
  • Neurotrophic effects of magnesium fructose 1, 6-diphosphate on cortical neurons.
Neurotrophic effects of magnesium fructose 1, 6-diphosphate on cortical neurons.
The International journal of neuroscience (2011-12-20)
Lian-Feng Lin, Xiao-Yan Xue, Min-Jing Liao, Fei Xiao, Ruo-Hua Lv, Huan-Min Luo
ABSTRACT
In this study, we evaluated the neurotrophic effects of magnesium fructose 1, 6-diphosphate (FDP-Mg) on cortical neurons. The results demonstrated that FDP-Mg promoted dendrite outgrowth and neuronal survival in a dose-dependent manner. In order to investigate the associated mechanisms, we determined adenosine triphosphate (ATP) levels and brain-derived neurotrophic factor (BDNF) mRNA expression in cortical neurons. Treatment with FDP-Mg significantly increased ATP levels and BDNF mRNA expression in cortical neurons. These data suggest that FDP-Mg can exert neurotrophic effects on cortical neurons. The increases in BDNF mRNA expression and cellular ATP levels are involved in the neurotrophic effects produced by FDP-Mg.
Sigma-Aldrich
D-Fructose 1,6-bisphosphate trisodium salt hydrate, ≥98% (TLC)