Merck
  • Protection/Deprotection of surface activity and its applications in the controlled release of liposomal contents.
Protection/Deprotection of surface activity and its applications in the controlled release of liposomal contents.
Langmuir : the ACS journal of surfaces and colloids (2012-02-07)
Xueshu Li, Yan Zhao
ABSTRACT
The micelles of two tripropargylammonium-functionalized cationic surfactants were cross-linked by a disulfide-containing diazido cross-linker in the presence of Cu(I) catalysts. With multiple residual alkyne groups on the surface, the resulting surface cross-linked micelles (SCMs) were postfunctionalized by reaction with 2-azidoethanol and an azido-terminated poly(ethylene glycol), respectively, via the alkyne-azide click reaction. The water-soluble nanoparticles obtained had low surface activity due to the buried hydrophobic tails. Cleavage of the disulfide cross-links by dithiothreitol (DTT) exposed the hydrophobic tails and resumed surface activity of the "caged" surfactants within 2 min after DTT addition. The controlled breakage of the SCMs was used to lower the surface tension of aqueous solutions and trigger the release of liposomal contents on demand.
Sigma-Aldrich
Tripropylamine, ≥98%
Pricing and availability is not currently available.