Merck
  • New cyclization mechanism for squalene: a ring-expansion step for the five-membered C-ring intermediate in hopene biosynthesis.
New cyclization mechanism for squalene: a ring-expansion step for the five-membered C-ring intermediate in hopene biosynthesis.
Bioscience, biotechnology, and biochemistry (2000-01-15)
T Hoshino, M Kouda, T Abe, S Ohashi
ABSTRACT
Three triterpenes having the 6/6/5-fused tri- and 6/6/6/5-fused tetracyclic skeletons were isolated from an incubation mixture of the mutated F601A enzyme, these products being in accordance with a Markovnikov closure. Successful trapping of the tricyclic cationic intermediate by using the squalene analog having a highly nucleophilic hydroxyl group leads us to propose that the ring expansion process of the 5-membered C-ring is involved in the squalene cyclization cascade.
Product Description
Supelco
Hop-22(29)-ene solution, 0.1 mg/mL in isooctane, analytical standard