Merck
The complete amino acid sequence of human Cu/Zn superoxide dismutase.
FEBS letters (1980-10-20)
D Barra, F Martini, J V Bannister, M E Schininà, G Rotilio, W H Bannister, F Bossa
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein