Merck
  • TLR-dependent activation stimuli associated with Th1 responses confer NK cell stimulatory capacity to mouse dendritic cells.
TLR-dependent activation stimuli associated with Th1 responses confer NK cell stimulatory capacity to mouse dendritic cells.
Journal of immunology (Baltimore, Md. : 1950) (2005-06-24)
Ivan Zanoni, Maria Foti, Paola Ricciardi-Castagnoli, Francesca Granucci
ABSTRACT
Dendritic cells (DCs) have an important role in the activation of NK cells that exert direct antitumor and antimicrobial effects and can influence the development of adaptive T cell responses. DCs acquire NK cell stimulatory capacity after exposure to various stimuli. In this study we investigated the nature of the stimuli that confer to DCs the NK cell-activating capacity. After exposure of DCs to TLR-dependent and -independent microbial stimuli and to nonmicrobial stimuli, we evaluated the ability of activated DCs to elicit IFN-gamma production from NK cells in vitro and to promote NK cell activation in vivo. We show in this study that only TLR-dependent microbial stimuli typically associated with Th1 responses confer to DCs the ability to activate NK cells, whereas stimuli associated with Th2 responses do not have this property.
Sigma-Aldrich
Pam3-Cys-OH, ≥98.0% (TLC)