Merck
  • N-{3-[(2-Ammonioethyl)amino]propyl}ethane-1,2-diaminium tris(trifluoromethanesulfonate): a salt of a folded triply protonated tetramine.
N-{3-[(2-Ammonioethyl)amino]propyl}ethane-1,2-diaminium tris(trifluoromethanesulfonate): a salt of a folded triply protonated tetramine.
Acta crystallographica. Section C, Crystal structure communications (2008-09-02)
Violetta Patroniak, Wanda Radecka-Paryzek, Maciej Kubicki
ABSTRACT
The structure of a trifluoromethanesulfonate salt of a nontypical triply protonated linear tetramine, C(7)H(23)N(4)(3+).3CF(3)SO(3)(-), with a layered crystal structure is presented. One N atom remains unprotonated. The conformation of the cation is enforced by intra- and intermolecular hydrogen bonds. The crystal structure is built of ca 10 A deep layers, within which cations and anions are hydrogen bonded. Each layer is only weakly bound to its neighbours. This study shows a rare example of an unsymmetrically protonated polyamine and the relation between the lack of protonation, intramolecular hydrogen bonding and the conformation of the cation.
Product Description
Supelco
Tetramethylenedisulfotetramine solution, 100 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®