Merck
  • Costunolide induces apoptosis through nuclear calcium2+ overload and DNA damage response in human prostate cancer.
Costunolide induces apoptosis through nuclear calcium2+ overload and DNA damage response in human prostate cancer.
The Journal of urology (2011-03-23)
Jui-Ling Hsu, Shiow-Lin Pan, Yunn-Fang Ho, Tsong-Long Hwang, Fan-Lu Kung, Jih-Hwa Guh
ABSTRACT
Costunolide is a natural sesquiterpene lactone. We elucidated what to our knowledge is a novel mechanism to highlight its potential in chemotherapy for prostate cancer, particularly androgen refractory prostate cancer. Several pharmacological and biochemical assays were used to characterize the apoptotic signaling pathways of costunolide (ChromaDex™) in prostate cancer cells. Costunolide showed effective antiproliferative activity against hormone dependent (LNCaP) and independent (PC-3 and DU-145) prostate cancer cells (ATCC®) by sulforhodamine B assay, clonogenic test and flow cytometric analysis of carboxyfluorescein succinimidyl ester labeling. In PC-3 cells data showed that costunolide induced a rapid overload of nuclear Ca(2+), DNA damage response and ATR phosphorylation. Costunolide induced G1-phase cell cycle arrest, which was supported by p21 up-regulation and its association with the cyclin dependent kinase 2/cyclin E complex. The association resulted in inhibition of the complex activity and inhibition of Rb phosphorylation. Costunolide mediated effects were substantially inhibited by glutathione, the reactive oxygen species scavenger and glutathione precursor N-acetylcysteine, and the Ca(2+) chelator BAPTA-AM other than the reactive oxygen species scavenger Trolox®. This indicated the crucial role of intracellular Ca(2+) mobilization and thiol depletion but not of reactive oxygen species production in apoptotic signaling. Data suggest that costunolide induces the depletion of intracellular thiols and overload of nuclear Ca(2+) that cause DNA damage and p21 up-regulation. The association of p21 with the cyclin dependent kinase 2/cyclin E complex blocks cyclin dependent kinase 2 activity and inhibits Rb phosphorylation, leading to G1 arrest of the cell cycle and subsequent apoptotic cell death in human prostate cancer cells.
Sigma-Aldrich
Costunolide, ≥97% (HPLC)