Merck
  • Vibrational spectroscopic, first-order hyperpolarizability and HOMO, LUMO studies of 1,2-dichloro-4-nitrobenzene based on Hartree-Fock and DFT calculations.
Vibrational spectroscopic, first-order hyperpolarizability and HOMO, LUMO studies of 1,2-dichloro-4-nitrobenzene based on Hartree-Fock and DFT calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-04-07)
M Arivazhagan, S Jeyavijayan
ABSTRACT
The Fourier-transform infrared spectrum of 1,2-dichloro-4-nitrobenzene (DCNB) was recorded in the region 4000-400cm(-1). The Fourier-transform Raman spectrum of DCNB was also recorded in the region 3500-50cm(-1). Quantum chemical calculations of energies, geometrical structure and vibrational wavenumbers of DCNB were carried out by ab initio HF and density functional theory (DFT/B3LYP) method with 6-31+G(d,p) basis set. The difference between the observed and scaled wavenumber values of most of the fundamentals is very small. The values of the total dipole moment (μ) and the first-order hyperpolarizability (β) of the investigated compound were computed using ab initio quantum mechanical calculations. The calculated results also show that the DCNB might have microscopic nonlinear optical (NLO) behavior with non-zero values. A detailed interpretation of the infrared and Raman spectra of DCNB is also reported based on total energy distribution (TED). The calculated HOMO and LUMO energies shows that charge transfer occur within the molecule. The theoretical FT-IR and FT-Raman spectra for the title compound have also been constructed.
Sigma-Aldrich
1,2-Dichloro-4-nitrobenzene, 99%
Pricing and availability is not currently available.