Merck
  • Pineal ultrastructure and indole profiles spanning the summer rise in arginine vasotocin immunoactivity.
Pineal ultrastructure and indole profiles spanning the summer rise in arginine vasotocin immunoactivity.
Endocrinology (1985-09-01)
J A McNulty, M M Prechel, T K Audhya, D Taylor, L Fox, T A Dombrowski, W H Simmons
ABSTRACT
A correlative radioimmunological-biochemical-ultrastructural study of the rat pineal gland was undertaken during the summer months when pineal arginine vasotocin (AVT) immunoactivity increases up to 200-fold. RIA confirmed a rapid rise in AVT activity during mid-August regardless of the time of day sampled. Pineal indoles were separated by HPLC and measured using electrochemical detection. Serotonin (5-HT) and 5-hydroxyindoleacetic acid levels were consistently elevated in daytime samples, and there was a significant trend for increased day and nighttime levels of 5-HT from July to September. Mid-dark levels of melatonin also exhibited a significant increase over the sample period. Nighttime levels of N-acetylserotonin mirrored fluctuations in 5-HT in the preceding photoperiod. Ultrastructural components implicated in peptide/protein and/or indole biosynthesis were quantified by stereological morphometry. The greatest amounts of rough endoplasmic reticulum stacks, lipid droplets, and annulate lamellae-like bodies coincided with peak AVT activity. Dense-cored vesicles and synaptic ribbons were consistently more frequent during the dark period. The number of dense-cored vesicles and nucleolar size tended to be greatest before and after the peak in AVT immunoactivity. These observations are consistent with the hypotheses that endoplasmic reticulum and lipid are functionally related to the synthesis and/or storage of peptide/protein factors and that numerical changes in synaptic ribbons and dense-cored vesicles are more closely related to day/night differences in indole metabolism.
Sigma-Aldrich
5-Methoxy-DL-tryptophan