Merck
  • Interlayer neurones in the rat superior colliculus: a tracer study using Dil/Di-ASP.
Interlayer neurones in the rat superior colliculus: a tracer study using Dil/Di-ASP.
Neuroreport (1994-01-12)
H Hilbig, A Schierwagen
ABSTRACT
Five different populations of interlayer neurones (ILNs) can be described after DiI/Di-ASP tracing in rat superior colliculus (SC). All of these labelled neurones preferentially lay in the rostro-medial part of the SC. Most of them are located in the stratum opticum and in the stratum griseum superficiale. Our results indicate that ILNs represent a minority of neurones in the superficial layers but may constitute a substantial population of neurones in the stratum opticum connecting the visual and the multimodal collicular layers.
Sigma-Aldrich
4-(4-Diethylaminostyryl)-1-methylpyridinium iodide, ≥97%