Merck
  • Description of two new alpha variants: Hb Canuts [alpha85(F6)Asp-->His (alpha1)] and Hb Ambroise Pare [alpha117(GH5)Phe-->Ile (alpha2)]; two new beta variants: Hb Beaujolais [beta84(EF8)Thr-->Asn] and Hb Monplaisir [beta147 (Tyr-Lys-Leu-Ala-Phe-Phe-Leu-Leu-Ser-Asn-Phe-Tyr-158-COOH)] and one new delta variant: Hb (A2)North Africa [delta59(E3)Lys-->Met].
Description of two new alpha variants: Hb Canuts [alpha85(F6)Asp-->His (alpha1)] and Hb Ambroise Pare [alpha117(GH5)Phe-->Ile (alpha2)]; two new beta variants: Hb Beaujolais [beta84(EF8)Thr-->Asn] and Hb Monplaisir [beta147 (Tyr-Lys-Leu-Ala-Phe-Phe-Leu-Leu-Ser-Asn-Phe-Tyr-158-COOH)] and one new delta variant: Hb (A2)North Africa [delta59(E3)Lys-->Met].
Hemoglobin (2009-08-07)
Philippe Joly, Philippe Lacan, Martine Bererd, Caroline Garcia, Isabelle Zanella-Cleon, Michel Becchi, Martine Aubry, Nicole Couprie, Alain Francina
ABSTRACT
We present here five new hemoglobin (Hb) variants which have been identified during routine Hb analysis before their genotypic characterization. Four of these result from a classical missense mutation: Hb Canuts [alpha85(F6)Asp-->His (alpha1)], Hb Ambroise Pare [alpha117(GH5)Phe-->Ile (alpha2)], Hb Beaujolais [beta84(EF8)Thr-->Asn] and HbA(2)-North Africa [delta59(E3)Lys-->Met]. The last one, Hb Monplaisir [beta147 (Tyr-Lys-Leu-Ala-Phe-Phe-Leu-Leu-Ser-Asn-Phe-Tyr-158-COOH)], results from a frameshift mutation at the stop codon of the beta-globin gene which leads to a modified C-terminal sequence in the beta-globin chain. None of these variants seem to have a particular clinical expression in the heterozygous state. The circumstances of the discovery of these five new Hb variants emphasize the fact that an association of techniques is necessary for a complete screening of Hb variants during routine Hb analysis. Globin chain separation by reversed phase liquid chromatography (RP-LC) appears to be the most relevant method.